Anti-OS9 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-519 NB100-519SS
NOVUNB100-519EA 554 EUR
NOVUNB100-519 NOVUNB100-519SS
Anti-OS9 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: OS9 (osteosarcoma amplified 9, endoplasmic reticulum lectin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR