Anti-NOD2 Mouse Monoclonal Antibody (DyLight® 488) [clone: 2D9]

Supplier: Novus Biologicals
NB100-524G
NOVUNB100-524GEA 724 EUR
NOVUNB100-524G
Anti-NOD2 Mouse Monoclonal Antibody (DyLight® 488) [clone: 2D9]
Antibodies
Type: Primary
Antigen: NOD2 (nucleotide-binding oligomerization domain containing 2)
Clonality: monoclonal
Clone: 2D9
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR