Anti-MBD1 Mouse monoclonal antibody [clone: 100B272.1]

Supplier: Novus Biologicals
NB100-56537SS
NOVUNB100-56537SSEA 260 EUR
NOVUNB100-56537SS
Anti-MBD1 Mouse monoclonal antibody [clone: 100B272.1]
Antibodies
The MBD1 Antibody (100B272.1) from Novus Biologicals is a mouse monoclonal antibody to MBD1. This antibody reacts with human, mouse. The MBD1 Antibody (100B272.1) has been validated for the following applications: Western Blot, Chromatin Immunoprecipitation, Immunoprecipitation.

Type: Primary
Antigen: MBD1
Clonality: Monoclonal
Clone: 100B272.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR