Anti-CrkRS Rabbit polyclonal antibody

Supplier: Novus Biologicals
NB100-87011
NOVUNB100-87011EA 774 EUR
NOVUNB100-87011
Anti-CrkRS Rabbit polyclonal antibody
Antibodies
The CrkRS Antibody from Novus Biologicals is a rabbit polyclonal antibody to CrkRS. This antibody reacts with human. The CrkRS Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: CrkRS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR