Anti-GPIHBP1 Rabbit Polyclonal Antibody (DyLight® 650)

Supplier: Novus Biologicals
NB110-41539C
NOVUNB110-41539CEA 722 EUR
NOVUNB110-41539C
Anti-GPIHBP1 Rabbit Polyclonal Antibody (DyLight® 650)
Antibodies
Type: Primary
Antigen: GPIHBP1 (glycosylphosphatidylinositol anchored high density lipoprotein binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR