Anti-IGF1R Mouse Monoclonal Antibody [clone: 3G5C1]

Supplier: Novus Biologicals
NB110-87052 NB110-87052SS
NOVUNB110-87052EA 623 EUR
NOVUNB110-87052 NOVUNB110-87052SS
Anti-IGF1R Mouse Monoclonal Antibody [clone: 3G5C1]
Antibodies
Mouse Monoclonal IGF1 Receptor Antibody (3G5C1). Tested Applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200- 1:1000

Type: Primary
Antigen: IGF1R
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR