Anti-CDKN2A Rat monoclonal antibody unconjugated [clone: 5-C3-1]

Supplier: Novus Biologicals
NB200-174 NB200-174SS
NOVUNB200-174EA 570 EUR
NOVUNB200-174 NOVUNB200-174SS
Anti-CDKN2A Rat monoclonal antibody unconjugated [clone: 5-C3-1]
Antibodies
Type: Primary
Antigen: CDKN2A (cyclin-dependent kinase inhibitor 2A)
Clonality: monoclonal
Clone: 5-C3-1
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR