Anti-SLC7A11 Rabbit Polyclonal Antibody (Biotin)

Supplier: Novus Biologicals
NB300-318B
NOVUNB300-318BEA 792 EUR
NOVUNB300-318B
Anti-SLC7A11 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: SLC7A11 (solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR