Anti-UCHL1 Rabbit Polyclonal Antibody (DyLight® 650)

Supplier: Novus Biologicals
NB300-676C
NOVUNB300-676CEA 620 EUR
NOVUNB300-676C
Anti-UCHL1 Rabbit Polyclonal Antibody (DyLight® 650)
Antibodies
Rabbit Polyclonal UCHL1 Antibody [DyLight 650]. Tested Applications: Western Blot, Immunohistochemistry. Tested Reactivity: Mouse.

Type: Primary
Antigen: UCHL1
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR