Anti-HPx2 Mouse Monoclonal Antibody [clone: HIC0-1C10]

Supplier: Novus Biologicals
NBP1-18952 NBP1-18952SS
NOVUNBP1-18952EA 568 EUR
NOVUNBP1-18952 NOVUNBP1-18952SS
Anti-HPx2 Mouse Monoclonal Antibody [clone: HIC0-1C10]
Antibodies
Type: Primary
Antigen: HPx2
Clonality: monoclonal
Clone: HIC0-1C10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR