Anti-BIRC3 Rabbit Polyclonal Antibody (DyLight® 488)

Supplier: Novus Biologicals
NBP1-27972G
NOVUNBP1-27972GEA 668 EUR
NOVUNBP1-27972G
Anti-BIRC3 Rabbit Polyclonal Antibody (DyLight® 488)
Antibodies
Type: Primary
Antigen: BIRC3 (baculoviral IAP repeat containing 3)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR