Anti-CD46 Mouse Monoclonal Antibody [clone: M177]

Supplier: Novus Biologicals
NBP1-51131
NOVUNBP1-51131EA 630 EUR
NOVUNBP1-51131
Anti-CD46 Mouse Monoclonal Antibody [clone: M177]
Antibodies
The CD46 Antibody (M177) from Novus Biologicals is a mouse monoclonal antibody to CD46. This antibody reacts with human. The CD46 Antibody (M177) has been validated for the following applications: Western Blot, Flow Cytometry, Immunoprecipitation, Functional, Immunoassay.

Type: Primary
Antigen: CD46
Clonality: Monoclonal
Clone: M177
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR