Anti-SNTB1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-59784
NOVUNBP1-59784EA 595 EUR
NOVUNBP1-59784
Anti-SNTB1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Syntrophin beta 1 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human.

Type: Primary
Antigen: Syntrophin beta 1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR