Anti-HSPB1 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Supplier: Novus Biologicals
NBP1-75477F
NOVUNBP1-75477FEA 636 EUR
NOVUNBP1-75477F
Anti-HSPB1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antibodies
Type: Primary
Antigen: HSPB1 (heat shock 27kDa protein 1)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR