Anti-RNF31 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-27290
NOVUNBP2-27290EA 434 EUR
NOVUNBP2-27290
Anti-RNF31 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RNF31 (ring finger protein 31)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR