Oil red O 0,5% in propylene glycol, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY

Synonyms: CI 26125, Sudan Red 5B, Solvent Red 27

M-Clarity™ quality level = MQ200

O1516-250ML O1516-500ML
O1516-250MLEA 381 EUR
O1516-250ML O1516-500ML
Oil red O 0,5% in propylene glycol, Sigma-Aldrich®
Oil Red O

Oil red O is a lysochrome (fat-soluble dye) diazo dye used for staining of neutral triglycerides and lipids on frozen sections and some lipoproteins on paraffin sections. In histology, a supersaturated solution of oil red O in isopropanol may be used to stain fat in tissue.

Formula: C₂₆H₂₄N₄O
MW: 408.51 g/mol
Storage Temperature: Room Temperature
MDL Number: MFCD00003898
CAS Number: 1320-06-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5db861df49353969f31bdacf110373f0af69e40e&vid=8127ea78128711195b44982188087011024fd2c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance
Dark Red, Sediment Free
Conforms
UV/VIS Absorbance
Lambda Max 506 to 520 nm
0.40 - 1.00
Lambda Max 506 - 520 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR