Orcein, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY


M-Clarity™ quality level = MQ100

O7380-10G O7380-5G
O7380-10GEA 200 EUR
O7380-10G O7380-5G
Orcein, Sigma-Aldrich®
Orcein
MDL Number: MFCD00062310
CAS Number: 1400-62-0
EINECS: 215-750-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR