Anti-DDT Mouse Monoclonal Antibody [clone: 2H3]

Supplier: OriGene
CF505658
ORIGCF505658EA 833 EUR
ORIGCF505658
Anti-DDT Mouse Monoclonal Antibody [clone: 2H3]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: DDT
Clonality: Monoclonal
Clone: 2H3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR