SLK Human shRNA Plasmid Kit (Locus ID 9748)
Supplier: OriGene
TL320620
ORIGTL320620EA 1410 EUR
ORIGTL320620
SLK Human shRNA Plasmid Kit (Locus ID 9748)
Vectors, Plasmids and Libraries
These shRNA constructs were designed against multiple splice variants at this gene locus.
  • Mediates apoptosis and actin stress fiber dissolution
Delivery information: Kit contains SLK - Human, 4 unique 29mer shRNA constructs in lentiviral GFP vector (Gene ID = 9748) 5 µg purified plasmid DNA per construct, 29-mer scrambled shRNA cassette in pGFP-C-shLenti Vector.