Protective armor for portable meters, Orion™ Star A series
Supplier: Thermo Orion
STARA-AR
ORIOSTARA-AREA 170 EUR
ORIOSTARA-AR
Protective armor for portable meters, Orion™ Star A series
Electrode Holders
This portable meter armor adds protection around the meter screen and includes a hand or belt strap that can be converted to built-in stand, pH electrode holder, conductivity and DO probe holder and pH electrode storage sleeve.
For use with Thermo Scientific™ Orion™ Star A series portable meters (Star A320, Star A220 and Star A120 series).
Delivery information: Supplied with hand strap and built-in stand, pH electrode storage sleeve, pH electrode holder, and conductivity and dissolved oxygen sensor holder.