Conductivity meter, handheld, Orion Star™ A322
Supplier: Thermo Orion
STARA3220 STARA3225 STARA3226
663-0216EA 1680 EUR
663-0216 663-0217 ORIOSTARA3226
Conductivity meter, handheld, Orion Star™ A322
Electrochemistry Meters Conductivity Meters
This portable conductivity meter is designed for a wide range of conductivity, total dissolved solids (TDS), salinity, resistivity and temperature testing and field applications. The meter is suitable for use in the most demanding locations thanks to its IP 67-rated housing.
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Measurement stability indicator and selectable read modes - Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific
More Product Information