Anti-IgG Goat Polyclonal Antibody (FITC)

Supplier: OZ BIOSCIENCES
IGG0100
OZBIIGG0100EA 107.5 EUR
OZBIIGG0100
Anti-IgG Goat Polyclonal Antibody (FITC)
Antibodies
Immunoglobulin (IgG) from goat serum has been labelled with the fluorescein isothiocyanate dye to produce a green-emitting antibody with a high fluorophore-to-protein ratio. The resulting antibody can be used as a positive control for intracellular antibody delivery with Ab-DeliverIN reagent.

Absorption maximum: 492 nm
Emission maximum: 520 nm
Extinction coefficient: 75000 M⁻¹cm⁻¹
Quantum yield: 0,79

Type: Control
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol IgG
Environmentally Preferable
Antigen name Immunoglobulin G
Antibody type Control
Clonality Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity
Host Goat
Isotype IgG
Form liquid
Storage buffer DPBS, pH 7,4
Storage temperature –20 °C
Shipping temperature RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR