Orthoboric acid Ph. Eur., Macron Fine Chemicals™

Supplier: Avantor

P144-33EA 0 EUR
P144-33 P144-42
Orthoboric acid Ph. Eur., Macron Fine Chemicals™
ortho-Boric acid
Formula: B(OH)₃
MDL Number: MFCD00011337
CAS Number: 10043-35-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR