Cleanroom shelf
Supplier: Palbam
MB06045-S
PALBMB06045-SEA 0 EUR
PALBMB06045-S
Cleanroom shelf
Furniture Shelves
Wall mounted mini shelf can be fixed anywhere. Fabrication material is stainless steel 304 with electropolished finish.
Order Now
SPECIFICATIONS
Environmentally Preferable