Tangential flow systems, Ultralab™
Supplier: Cytiva (Formerly Pall Lab)
FS016X75 FS017X70
PALLFS016X75EA 13470 EUR
PALLFS016X75 PALLFS017X70
Tangential flow systems, Ultralab™
Filters Tangential Flow Systems
These systems are designed for use in diafiltration procedures which are faster and more efficient than dialysis for buffer exchange and desalting, and for concentration of dilute sample volumes of up to five litres.
  • Feed pressure gauge monitors sample processing
  • Three-way sampling valve allows maximum sample recovery without decanting
  • Peristaltic pump action minimises shear of sensitive molecules, eliminates air in the system and reduces foaming
  • A variable speed controller allows precise adjustment of crossflow during sample processing