Syringes for GC autosystem
Supplier: PerkinElmer
00230111 00230113 00230177 00230178 00230523 N6101252 N6101253 N6101390 N6101760 N9302203 N9302204 N9302210 N9302211 N9302212 N9302213 N9302214 N9302215 N9302231 N9302240 N9302241 N9302243 N9302244 N9302245 N9302250 N9302251 N9302253 N9302257 N9302242 N6101380 N9302266 N6101251 N9302235 N9302247 N9302256 N9302259 N9302262 N9302265 09904822 09904823 09904941
PERC00230111EA 374 EUR
PERC00230111 PERC00230113 PERC00230177 PERC00230178 PERC00230523 PERCN6101252 PERCN6101253 PERCN6101390 PERCN6101760 PERCN9302203 PERCN9302204 PERCN9302210 PERCN9302211 PERCN9302212 PERCN9302213 PERCN9302214 PERCN9302215 PERCN9302231 PERCN9302240 PERCN9302241 PERCN9302243 PERCN9302244 PERCN9302245 PERCN9302250 PERCN9302251 PERCN9302253 PERCN9302257 PERCN9302242 PERCN6101380 PERCN9302266 PERCN6101251 PERCN9302235 PERCN9302247 PERCN9302256 PERCN9302259 PERCN9302262 PERCN9302265 PERC09904822 PERC09904823 PERC09904941
Syringes for GC autosystem
Syringes Syringes for Chromatography
Syringes must ensure a tight and secure fit in the injection port and withstand repeated injection cycles without bending or breaking.
  • Can withstand repeatable injections
  • Variety of fixed and removable needles
  • Inspected to ensure accuracy and performance
Autosampler syringes are available in 0,5, 5 and 50 μl capacities. For routine analyses, the metal plunger in barrel with PTFE-tipped seal is the standard syringe with each GC instrument.
Ultra-micro volume syringes are recommended for liquid sample injections of less than 5 μl for gasMore Product Information