Guanidinium hydrochloride

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

24110
PIER24110EA 210 EUR
PIER24110
Guanidinium hydrochloride
Guanidinium hydrochloride
Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Melting Pt: 185
Density: 1.354 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR