1-Step™, Pierce™ Chloronaphthol
Supplier: Thermo Fisher Scientific
34012
PIER34012EA 138 EUR
PIER34012
1-Step™, Pierce™ Chloronaphthol
Electrophoresis Reagents Western Blotting Reagents
Chloronaphthol (4-CN) is a peroxidase substrate used for chromogenic detection of HRP in immunoblotting and immunohistochemical applications. This product is less sensitive than other precipitating substrates, but the distinct blue to blue-purple precipitate photographs well and can be useful in double-staining applications. The coloured product is soluble in alcohol and xylenes, but is not soluble in aqueous buffers. The 1-Step Chloronaphthol is supplied ready to use.