Freund's incomplete adjuvant, Imject™
Supplier: Thermo Fisher Scientific
77145
PIER77145EA 133 EUR
PIER77145
Freund's incomplete adjuvant, Imject™
Protein Expression Protein Expression Reagents
Imject™ Freund's Incomplete Adjuvant (FIA) is a popular non specific stimulator of the immune response for use in antibody production immunisation protocols.
  • Enhances immune response in booster injections
  • Typically used for subsequent boosts after initial injection with FCA
  • Water-in-oil emulsion without Mycobacterium
Delivery information: Supplied ready to use in 10 ml ampoules.
Order Now
SPECIFICATIONS
Environmentally Preferable