Micrococcal nuclease solution (≥100 U/µl)
Supplier: Thermo Fisher Scientific
88216
PIER88216EA 197 EUR
PIER88216
Micrococcal nuclease solution (≥100 U/µl)
Protein Extraction Protein Extraction Reagents
This Micrococcal Nuclease (MNase) is suitable for removing nucleic acids from cell lysates, releasing chromatin-bound proteins and shearing chromatin for use in chromatin immunoprecipitation (ChIP) experiments.
  • Enzyme digests nucleic acids (DNA and RNA)
  • Effective for double-stranded, single-stranded, circular and linear nucleic acids
  • Active in neutral to alkaline buffer conditions containing divalent calcium ions
  • Highest activity is toward single-stranded nucleic acid substrates with preference for AT or AU rich regions
Order Now
SPECIFICATIONS
Environmentally Preferable