Microdialysis plates, 96-well, Pierce™
Supplier: Thermo Fisher Scientific
88260 88262
PIER88260EA 610 EUR
PIER88260 PIER88262
Microdialysis plates, 96-well, Pierce™
Microplates Deep Well Microplates
Pierce™ 96-well microdialysis plates are automation-compatible microplate dialysis devices suitable for rapidly processing 1 to 96 samples with volumes 10 to 100 µl.
  • High yield, > 90% protein recovery after dialysis
  • Dialysis completed in 2 to 4 hours
  • Sample loading and retrieval can be performed using a standard pipette
  • Detachable 8-device strips, scalable from 1 to 96 samples
  • Plate format conforms to the SBS microplate standard
Pierce™ 96-well microdialysis plates are suitable for applications such as protein and DNA desalting/buffer exchange,More Product Information