Dapsone

Supplier: LGC Standards PROMOCHEM
Warning

BP102
PROOBP102EA 378 EUR
PROOBP102
Dapsone
Dapsone
Formula: C₁₂H₁₂N₂O₂S
MDL Number: MFCD00007887
CAS Number: 80-08-0
EINECS: 201-248-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR