Prednisone

Supplier: LGC Standards PROMOCHEM

BP553
PROOBP553EA 324 EUR
PROOBP553
Prednisone
Prednisone
Formula: C₂₁H₂₆O₅
MW: 358.43 g/mol
Melting Pt: 230…232 °C
MDL Number: MFCD00003608
CAS Number: 53-03-2
EINECS: 200-160-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR