Diphenyl phthalate

Supplier: LGC Standards PROMOCHEM
Warning

Synonyms: Phthalic acid diphenyl ester, Phenyl phthalate

CHE147
PROOCHE147EA 260 EUR
PROOCHE147
Diphenyl phthalate
Diphenyl phthalate
Formula: C₂₀H₁₄O₄
MW: 318.33 g/mol
Boiling Pt: 405
Melting Pt: 74…76 °C
Density: 1.313 g/cm³ (20 °C)
MDL Number: MFCD00003038
CAS Number: 84-62-8
EINECS: 201-546-4
Merck Index: 12,07460

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR