Ethylene glycol

Supplier: LGC Standards PROMOCHEM
Warning

Synonyms: Ethane-1,2-diol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, Glycol

CHE155
PROOCHE155EA 260 EUR
PROOCHE155
Ethylene glycol
Ethylene glycol
Formula: HOCH₂CH₂OH
MW: 62.07 g/mol
Boiling Pt: 196…198 °C (1013 hPa)
Melting Pt: –13 °C
Density: 1.115 g/cm³ (20 °C)
Flash Pt: 116
Storage Temperature: Refrigerator
MDL Number: MFCD00002885
CAS Number: 107-21-1
EINECS: 203-473-3
REACH: 01-2119456816-28
Merck Index: 13,03832
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d1c97392851ba184f445f6ee912f84631f98d9&vid=2002300f174929a030d66d9f1394eb6bee89b7dc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR