1-Decanol

Supplier: LGC Standards PROMOCHEM
Warning

Synonyms: 1-Hydroxy decane, 1-Decyl alcohol, Capric alcohol, Decan-1-ol, Nonyl carbinol, Decyl alcohol

CHE172
PROOCHE172EA 260 EUR
PROOCHE172
1-Decanol
1-Decanol
Formula: H₃C(CH₂)₈CH₂OH
MW: 158.28 g/mol
Boiling Pt: 230
Melting Pt: 7
Density: 0.82634 g/cm³ (20 °C)
Flash Pt: 82
Storage Temperature: Ambient
MDL Number: MFCD00004747
CAS Number: 112-30-1
EINECS: 203-956-9
UN: 3082
ADR: 9 III
Merck Index: 13,02875

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR