2,2',3,3',4,6,6'-Heptachlorobiphenyl (PCB No. 176)

Supplier: LGC Standards PROMOCHEM

CIL-PCB-176-C
PROOCIL-PCB-176-CEA 860 EUR
PROOCIL-PCB-176-C
2,2',3,3',4,6,6'-Heptachlorobiphenyl (PCB No. 176)
2,2',3,3',4,6,6'-Heptachlorobiphenyl (PCB No. 176)
Formula: C₁₂H₃Cl₇
MW: 395.32 g/mol
CAS Number: 52663-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR