2,2',6-Trichlorobiphenyl (PCB No. 19)

Supplier: LGC Standards PROMOCHEM

CIL-PCB-19-C
PROOCIL-PCB-19-CEA 860 EUR
PROOCIL-PCB-19-C
2,2',6-Trichlorobiphenyl (PCB No. 19)
2,2',6-Trichlorobiphenyl (PCB No. 19)
Formula: C₁₂H₇Cl₃
MW: 257.55 g/mol
CAS Number: 38444-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR