2,3,3',4',5,5',6-Heptachlorobiphenyl (PCB No. 193)

Supplier: LGC Standards PROMOCHEM

CIL-PCB-193-C
PROOCIL-PCB-193-CEA 895 EUR
PROOCIL-PCB-193-C
2,3,3',4',5,5',6-Heptachlorobiphenyl (PCB No. 193)
2,3,3',4',5,5',6-Heptachlorobiphenyl (PCB No. 193)
Formula: C₁₂H₃Cl₇
MW: 395.32 g/mol
CAS Number: 69782-91-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR