1,2-Dichloropropane-[D₆] (98% D)

Supplier: LGC Standards PROMOCHEM
Danger

CILDLM19370.1
PROOCILDLM19370.1EA 266 EUR
PROOCILDLM19370.1
1,2-Dichloropropane-[D₆] (98% D)
1,2-Dichloropropane-[D6]
Formula: ClD₂CCD₂CD₂Cl
Storage Temperature: Ambient
CAS Number: 93952-08-0
UN: 1279
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR