2-Chloro-9,10-anthraquinone

Supplier: LGC Standards PROMOCHEM
Warning

Synonyms: 2-Chloroanthraquinone

ERM-FC022
PROOERM-FC022EA 364 EUR
PROOERM-FC022
2-Chloro-9,10-anthraquinone
2-Chloro-9,10-anthraquinone
Formula: C₁₄H₇ClO₂
MW: 242.66 g/mol
Boiling Pt: 292 °C (30 mmHg)
Melting Pt: 207…210 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00001227
CAS Number: 131-09-9
EINECS: 205-010-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR