4-Nitrotoluene

Supplier: LGC Standards PROMOCHEM
Danger

ERM-FC029
PROOERM-FC029EA 364 EUR
PROOERM-FC029
4-Nitrotoluene
4-Nitrotoluene
Formula: C₇H₇NO₂
Boiling Pt: 238,3 °C (1013 hPa)
Melting Pt: 51 °C
Density: 1,39 g/cm³ (20 °C)
Flash Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD00007366
CAS Number: 99-99-0
EINECS: 202-808-0
UN: 3446
ADR: 6.1,II
Merck Index: 12,06748

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR