Riboflavine (vitamin B2)

Supplier: LGC Standards PROMOCHEM

Synonyms: Vitamin B2, Lactoflavin, 7,8-Dimethyl-10-((2S,3S,4R)-2,3,4,5-tetrahydroxypentyl)benzo[g]pteridine-2,4(3H,10H)-dione, (-)-Riboflavin, Vitamine G

ICRS0408
PROOICRS0408EA 288 EUR
PROOICRS0408
Riboflavine (vitamin B2)
Riboflavine (Vitamin B2)
Formula: C₁₇H₂₀N₄O₆
MW: 376,37 g/mol
Melting Pt: 280 °C
Storage Temperature: Ambient
MDL Number: MFCD00005022
CAS Number: 83-88-5
EINECS: 201-507-1
Merck Index: 12,08367

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR