3-Acetylbenzophenone

Supplier: LGC Standards PROMOCHEM

Synonyms: 1-(3-Benzoylphenyl)ethan-1-one, 1-(3-Benzoylphenyl)ethanone

MM0001.01
PROOMM0001.01EA 1290 EUR
PROOMM0001.01
3-Acetylbenzophenone
3-Acetylbenzophenone
Formula: C₁₅H₁₂O₂
MW: 224.2546 g/mol
MDL Number: MFCD01662280
CAS Number: 66067-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR