(±)-3-Chloro-1,2-propanediol

Supplier: LGC Standards PROMOCHEM

MM0027.22
PROOMM0027.22EA 403 EUR
PROOMM0027.22
(±)-3-Chloro-1,2-propanediol
(±)-3-Chloro-1,2-propanediol
Formula: C₃H₇ClO₂
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: –40 °C
Density: 1,32 g/cm³ (20 °C)
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00004712
CAS Number: 96-24-2
EINECS: 202-492-4
UN: 2689
ADR: 6.1,III
Merck Index: 13,02164

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR