Cloxiquina

Supplier: LGC Standards PROMOCHEM
Warning

Synonyms: 5-Chloro-8-quinolinol, 5-Chloroquinolin-8-ol, 5-Chloro-8-hydroxyquinoline

MM0391.00
PROOMM0391.00EA 329 EUR
PROOMM0391.00
Cloxiquina
Cloxiquina
Formula: C₉H₆ClNO
MW: 179.61 g/mol
Boiling Pt: 349
Melting Pt: 125
Storage Temperature: Ambient
MDL Number: MFCD00006788
CAS Number: 130-16-5
EINECS: 204-978-1
Merck Index: 12,02483

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR