L(+)-Potassium hydrogen tartrate

Supplier: LGC Standards PROMOCHEM

NIST-188
PROONIST-188EA 1220 EUR
PROONIST-188
L(+)-Potassium hydrogen tartrate
L(+)-Potassium hydrogen tartrate
Formula: KOOC(CHOH)₂COOH
Melting Pt: 250 °C
Density: 1,943 g/cm³ (20 °C)
MDL Number: MFCD00065392
CAS Number: 868-14-4
EINECS: 212-769-1
Merck Index: 13,07697
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=355f72f319f5589f12d60c6dfffecdec31cee083&vid=4a750ad5d4a91fd4c2885832ee80b7b64cdcc42a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR