Clofenotane

Supplier: LGC Standards PROMOCHEM
Danger

NMIAP1309
PROONMIAP1309EA 317 EUR
PROONMIAP1309
Clofenotane
Clofenotane
Formula: C₁₄H₉Cl₅
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: ∼89 °C
Density: ∼1,5 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 50-29-3
EINECS: 200-024-3
UN: 2761
ADR: 6.1,III
Merck Index: 12,02898

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR