Sodium metabisulphite

Supplier: LGC Standards PROMOCHEM
Danger

Synonyms: Disodium disulphite, di-Sodium disulphite, Sodium pyrosulphite, Disodiumdisulphite, Sodium disulphite

TRC-S66706010G
PROOTRC-S66706010GEA 294 EUR
PROOTRC-S66706010G
Sodium metabisulphite
Sodium metabisulphite

Caution: Use of water spray when fighting fire may be inefficient.

Formula: Na₂S₂O₅
MW: 190.11 g/mol
Melting Pt: 150
MDL Number: MFCD00167602
CAS Number: 7681-57-4
Merck Index: 12,08784

Order Now

Specification Test Results

Appearance White Solid
MS Conforms
Sodium Content 24.6%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR