Silver standard solution, for ICP, in HNO₃, CRM, VHG
Supplier: LGC Standards PROMOCHEM
VHG-PAGN-100 VHG-PAGN-500 VHG-TAGN-100 VHG-TAGN-500 VHG-LAGN-100
PROOVHG-PAGN-100EA 63.5 EUR
PROOVHG-PAGN-100 PROOVHG-PAGN-500 PROOVHG-TAGN-100 PROOVHG-TAGN-500 PROOVHG-LAGN-100
Silver standard solution, for ICP, in HNO₃, CRM, VHG
Standards Cationic Standards Silver Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.